tert-Butyl-7-(benzyloxy)-3-formyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

071719-1G 071719-5G 071719-10G
102520-940EA 355.38 USD
102520-940 102520-942 102520-944
tert-Butyl-7-(benzyloxy)-3-formyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-7-(benzyloxy)-3-formyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
CAS Number: 914348-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR