tert-Butyl-6-(benzyloxy)-3-formyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6-(benzyloxy)-3-formyl-1H-indole-1-carboxylate

071717-1G 071717-5G 071717-10G
102520-928EA 355.38 USD
102520-928 102520-930 102520-932
tert-Butyl-6-(benzyloxy)-3-formyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-6-(benzyloxy)-3-formyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.3958 g/mol
MDL Number: MFCD05864702
CAS Number: 630110-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR