6-Benzyloxyindole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

071716-1G 071716-5G 071716-10G
102520-922EA 355.38 USD
102520-922 102520-924 102520-926
6-Benzyloxyindole-3-carbaldehyde ≥95%
6-Benzyloxyindole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₃NO₂
MW: 251.2800 g/mol
MDL Number: MFCD00056931
CAS Number: 92855-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR