tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Benzyloxy)-3-formyl-1H-indole, N-BOC protected

071715-1G 071715-5G 071715-10G
102520-916EA 355.38 USD
102520-916 102520-918 102520-920
tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
CAS Number: 914348-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR