1-Boc-4-benzyloxy-3-formylindole ≥95%

Supplier: Matrix Scientific

071714-1G 071714-5G 071714-10G
102520-910EA 355.38 USD
102520-910 102520-912 102520-914
1-Boc-4-benzyloxy-3-formylindole ≥95%
1-Boc-4-benzyloxy-3-formylindole

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
CAS Number: 404888-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR