4-Benzyloxyindole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

071713-5G 071713-10G
102520-906EA 783.08 USD
102520-906 102520-908
4-Benzyloxyindole-3-carbaldehyde ≥95%
4-Benzyloxyindole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₃NO₂
MW: 251.28 g/mol
MDL Number: MFCD00152002
CAS Number: 7042-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR