7-Nitroindole-3-carboxyaldehyde ≥95%
Supplier: Matrix Scientific
071712-1G
071712-5G
071712-10G
102520-900EA
355.38
USD
102520-900
102520-902
102520-904
7-Nitroindole-3-carboxyaldehyde ≥95%
7-Nitroindole-3-carboxyaldehyde
Hazard: Irritant
Formula:
C₉H₆N₂O₃ MW: 190.16 g/mol |
MDL Number:
MFCD01074519 CAS Number: 10553-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...