tert-Butyl-3-formyl-7-nitro-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

071711-1G 071711-5G 071711-10G
102520-894EA 355.38 USD
102520-894 102520-896 102520-898
tert-Butyl-3-formyl-7-nitro-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-formyl-7-nitro-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₅
CAS Number: 914348-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR