tert-Butyl-3-formyl-6-methyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methylindole-3-carboxaldehyde, N-BOC protected

071708-1G 071708-5G 071708-10G
102520-878EA 355.38 USD
102520-878 102520-880 102833-506
tert-Butyl-3-formyl-6-methyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-formyl-6-methyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₇NO₃
CAS Number: 914348-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR