tert-Butyl-3-formyl-5-methyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methylindole-3-carboxaldehyde, N-BOC protected

071707-1G 071707-5G 071707-10G
102520-872EA 355.38 USD
102520-872 102520-874 102520-876
tert-Butyl-3-formyl-5-methyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-formyl-5-methyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₇NO₃
CAS Number: 914348-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR