1-Boc-4-bromo-3-formylindole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-4-Bromo-3-formylindole, 4-Bromo-1H-indole-3-carboxaldehyde, N-BOC protected

071706-1G 071706-5G 071706-10G
102520-866EA 355.38 USD
102520-866 102520-868 102520-870
1-Boc-4-bromo-3-formylindole ≥95%
1-Boc-4-bromo-3-formylindole

Hazard: Irritant

Formula: C₁₄H₁₄BrNO₃
MW: 324.1699 g/mol
MDL Number: MFCD05864694
CAS Number: 303041-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR