tert-Butyl-5-cyano-3-formyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Cyano-1H-indole-3-carboxaldehyde, N-BOC protected

071704-1G 071704-5G 071704-10G
102520-854EA 160 USD
102520-854 102520-856 102520-858
tert-Butyl-5-cyano-3-formyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-5-cyano-3-formyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₃
CAS Number: 914348-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR