tert-Butyl-5-bromo-3-formyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5-bromo-3-formyl-1H-indole-1-carboxylate, 5-Bromo-1H-indole-3-carboxaldehyde, N-BOC protected

071703-5G 071703-10G 071703-100G
102520-848EA 320 USD
102520-848 102520-850 102520-852
tert-Butyl-5-bromo-3-formyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-5-bromo-3-formyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄BrNO₃
MW: 324.1700 g/mol
MDL Number: MFCD05864691
CAS Number: 325800-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR