tert-Butyl-3-(4-chlorophenyl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)piperazine, N1-BOC protected, tert-Butyl 3-(4-chlorophenyl)piperazine-1-carboxylate

071701-1G 071701-5G
102520-838EA 640 USD
102520-838 102520-840
tert-Butyl-3-(4-chlorophenyl)piperazine-1-carboxylate ≥95%
tert-Butyl-3-(4-chlorophenyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₁ClN₂O₂
MW: 296.7924 g/mol
MDL Number: MFCD03840084
CAS Number: 886767-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR