4-(1H-1,2,4-Triazol-1-yl)aniline ≥95%

Supplier: Matrix Scientific

071700-1G 071700-5G 071700-10G
102520-832EA 178.46 USD
102520-832 102520-834 102520-836
4-(1H-1,2,4-Triazol-1-yl)aniline ≥95%
4-(1H-1,2,4-Triazol-1-yl)aniline

Hazard: Irritant

Formula: C₈H₈N₄
MDL Number: MFCD00171470
CAS Number: 6523-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR