2-(2,4-Dichloro-5-fluorophenyl)piperazine ≥95%

Supplier: Matrix Scientific

071699-1G 071699-5G 071699-10G
102520-826EA 640 USD
102520-826 102520-828 102520-830
2-(2,4-Dichloro-5-fluorophenyl)piperazine ≥95%
2-(2,4-Dichloro-5-fluorophenyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₁Cl₂FN₂
CAS Number: 914348-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR