2-(2,5-Dichlorophenyl)piperazine ≥95%

Supplier: Matrix Scientific

071698-1G 071698-5G 071698-10G
102520-820EA 640 USD
102520-820 102520-822 102520-824
2-(2,5-Dichlorophenyl)piperazine ≥95%
2-(2,5-Dichlorophenyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₂Cl₂N₂
CAS Number: 914348-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR