2-(3-Nitrophenyl)piperazine ≥95%

Supplier: Matrix Scientific

071694-1G 071694-5G 071694-10G
102520-796EA 915.38 USD
102520-796 102520-798 102520-800
2-(3-Nitrophenyl)piperazine ≥95%
2-(3-Nitrophenyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₂
CAS Number: 65709-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR