2-(3,5-Dimethoxyphenyl)piperazine ≥95%

Supplier: Matrix Scientific

071691-1G 071691-5G 071691-10G
102520-778EA 640 USD
102520-778 102520-780 102520-782
2-(3,5-Dimethoxyphenyl)piperazine ≥95%
2-(3,5-Dimethoxyphenyl)piperazine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
CAS Number: 910444-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR