2-(5-Bromothiophen-2-yl)piperazine ≥95%

Supplier: Matrix Scientific

071690-1G 071690-5G 071690-10G
102520-772EA 640 USD
102520-772 102520-774 102520-776
2-(5-Bromothiophen-2-yl)piperazine ≥95%
2-(5-Bromothiophen-2-yl)piperazine

Hazard: Irritant

Formula: C₈H₁₁BrN₂S
CAS Number: 111760-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR