2-(4-Trifluoromethylphenyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Trifluoromethylphenyl)piperazine dihydrochloride

071689-1G 071689-5G 071689-10G
102520-766EA 301.54 USD
102520-766 102520-768 102520-770
2-(4-Trifluoromethylphenyl)piperazine ≥95%
2-(4-Trifluoromethylphenyl)piperazine

Hazard: Corrosive

Formula: C₁₁H₁₃F₃N₂
MW: 230.2300 g/mol
MDL Number: MFCD05864678
CAS Number: 185110-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR