2-(4-Ethylphenyl)piperazine ≥95%

Supplier: Matrix Scientific

071688-1G 071688-5G 071688-10G
102520-760EA 640 USD
102520-760 102520-762 102520-764
2-(4-Ethylphenyl)piperazine ≥95%
2-(4-Ethylphenyl)piperazine

Hazard: Irritant

Formula: C₁₂H₁₈N₂
CAS Number: 910444-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR