2-(4-Fluorophenyl)piperazine ≥95%

Supplier: Matrix Scientific

071686-1G 071686-5G 071686-10G
102520-748EA 355.38 USD
102520-748 102520-750 102520-752
2-(4-Fluorophenyl)piperazine ≥95%
2-(4-Fluorophenyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₃FN₂
CAS Number: 65709-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR