2-(4-Bromophenyl)piperazine ≥95%

Supplier: Matrix Scientific

071685-1G 071685-5G 071685-10G
102520-742EA 533.85 USD
102520-742 102520-744 102520-746
2-(4-Bromophenyl)piperazine ≥95%
2-(4-Bromophenyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂
CAS Number: 105242-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR