2-(4-Nitro-2-(trifluoromethyl)phenyl)-1,2,3,4-tetrahydroisoquinoline ≥95%

Supplier: Matrix Scientific

071684-1G 071684-5G 071684-10G
102520-736EA 355.38 USD
102520-736 102520-738 102520-740
2-(4-Nitro-2-(trifluoromethyl)phenyl)-1,2,3,4-tetrahydroisoquinoline ≥95%
2-(4-Nitro-2-(trifluoromethyl)phenyl)-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₁₆H₁₃F₃N₂O₂
CAS Number: 914348-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR