4-(4-Methylpiperidin-1-yl)-3-(trifluoromethyl)aniline dihydrochloride ≥95%

Supplier: Matrix Scientific

071683-1G 071683-5G 071683-10G
102520-730EA 355.38 USD
102520-730 102520-732 102520-734
4-(4-Methylpiperidin-1-yl)-3-(trifluoromethyl)aniline dihydrochloride ≥95%
4-(4-Methylpiperidin-1-yl)-3-(trifluoromethyl)aniline dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂F₃N₂
CAS Number: 1189494-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR