1-(4-Nitro-2-(trifluoromethyl)phenyl)piperazine ≥95%

Supplier: Matrix Scientific

071682-1G 071682-5G
102520-726EA 213.85 USD
102520-726 102520-728
1-(4-Nitro-2-(trifluoromethyl)phenyl)piperazine ≥95%
1-(4-Nitro-2-(trifluoromethyl)phenyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₂F₃N₃O₂
CAS Number: 381242-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR