1-Boc-4-(4-formylphenyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-4-(4-Formylphenyl)piperazine, 4-Piperazin-1-ylbenzaldehyde, N4-BOC protected

071675-1G 071675-5G
102520-688EA 160 USD
102520-688 102520-690
1-Boc-4-(4-formylphenyl)piperazine ≥95%
1-Boc-4-(4-formylphenyl)piperazine

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₃
MW: 290.3600 g/mol
MDL Number: MFCD05864663
CAS Number: 197638-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR