1-Methyl-3,4-bis(3-indolyl)maleimide ≥95%
Supplier: Matrix Scientific
Synonyms:
3,4-Di(1H-indol-3-yl)-1-methyl-1H-pyrrole-2,5-dione, Bisindolylmaleimide V
071672-250MG
071672-1G
102520-672EA
710.77
USD
102520-672
102520-674
1-Methyl-3,4-bis(3-indolyl)maleimide ≥95%
1-Methyl-3,4-bis(3-indolyl)maleimide
Hazard: Irritant
Formula:
C₂₁H₁₅N₃O₂ MW: 341.37 g/mol |
MDL Number:
MFCD00236433 CAS Number: 113963-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...