3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione, 3,4-Bis(3-indolyl)maleimide, Bisindolylmaleimide IV, Arcyriarubin A

071671-250MG 071671-1G
102520-668EA 710.77 USD
102520-668 102520-670
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥95%
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C20H13N3O2
MW: 327.3361 g/mol
MDL Number: MFCD00236432
CAS Number: 119139-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR