2-(4-Trifluoromethylphenyl)thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

071670-1G 071670-5G
102520-664EA 710.77 USD
102520-664 102520-666
2-(4-Trifluoromethylphenyl)thiazole-5-carbaldehyde ≥95%
2-(4-Trifluoromethylphenyl)thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₆F₃NOS
CAS Number: 447406-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR