2-Amino-4-chlorothiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

071665-1G 071665-5G 071665-10G
102520-634EA 184.62 USD
102520-634 102520-636 102520-638
2-Amino-4-chlorothiazole-5-carbaldehyde ≥95%
2-Amino-4-chlorothiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₄H₃ClN₂OS
MW: 162.5974 g/mol
MDL Number: MFCD00204993
CAS Number: 76874-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR