2-(Benzo[d]oxazol-2-ylthio)-4-chlorothiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

071664-1G 071664-5G
102520-630EA 533.85 USD
102520-630 102520-632
2-(Benzo[d]oxazol-2-ylthio)-4-chlorothiazole-5-carbaldehyde ≥95%

Hazard: Irritant

Formula: C₁₁H₅ClN₂O₂S₂
CAS Number: 914348-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR