4-Chloro-2-(4-hydroxypiperidin-1-yl)thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

071663-1G 071663-5G
102520-626EA 533.85 USD
102520-626 102520-628
4-Chloro-2-(4-hydroxypiperidin-1-yl)thiazole-5-carbaldehyde ≥95%
4-Chloro-2-(4-hydroxypiperidin-1-yl)thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂S
CAS Number: 883107-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR