Methyl-1-(4-chloro-5-formylthiazol-2-yl)piperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

071662-1G 071662-5G
102520-622EA 533.85 USD
102520-622 102520-624
Methyl-1-(4-chloro-5-formylthiazol-2-yl)piperidine-4-carboxylate ≥95%
Methyl-1-(4-chloro-5-formylthiazol-2-yl)piperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃S
CAS Number: 914348-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR