4-Chloro-2-(1-piperidino)-5-thiazolecarboxaldehyde ≥95%

Supplier: Matrix Scientific

071661-1G 071661-5G
102520-618EA 533.85 USD
102520-618 102520-620
4-Chloro-2-(1-piperidino)-5-thiazolecarboxaldehyde ≥95%
4-Chloro-2-(1-piperidino)-5-thiazolecarboxaldehyde

Hazard: Irritant

Formula: C₉H₁₁ClN₂OS
CAS Number: 139670-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR