4-Chloro-2-(tert-butylamino)-5-thiazolecarboxaldehyde ≥95%

Supplier: Matrix Scientific

071659-1G 071659-5G
102520-608EA 640 USD
102520-608 102520-610
4-Chloro-2-(tert-butylamino)-5-thiazolecarboxaldehyde ≥95%
4-Chloro-2-(tert-butylamino)-5-thiazolecarboxaldehyde

Hazard: Irritant

Formula: C₈H₁₁ClN₂OS
CAS Number: 199851-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR