2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: (S,S)-Ph-pybox, (-)-2,6-Bis[(4S)-4-phenyl-2-oxazolin-2-yl]pyridine

071658-1G 071658-5G 071658-10G
102520-602EA 355.38 USD
102520-602 102520-604 102520-606
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

Hazard: Irritant

Formula: C₂₃H₁₉N₃O₂
MDL Number: MFCD01863586
CAS Number: 174500-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR