2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: (R,R)-Ph-pybox, (+)-2,6-Bis[(4R)-4-phenyl-2-oxazolin-2-yl]pyridine

071657-1G 071657-5G 071657-10G
102520-596EA 533.85 USD
102520-596 102520-598 102520-600
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

Hazard: Irritant

Formula: C₂₃H₁₉N₃O₂
MDL Number: MFCD01863585
CAS Number: 128249-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR