Ethyl-4-amino-3-fluorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-amino-3-fluorobenzoate

071656-1G 071656-5G 071656-10G
102520-590EA 427.69 USD
102520-590 102520-592 102520-594
Ethyl-4-amino-3-fluorobenzoate ≥95%
Ethyl 4-amino-3-fluorobenzoate

Hazard: Irritant

Formula: C₉H₁₀FNO₂
MW: 183.1796 g/mol
MDL Number: MFCD07782043
CAS Number: 73792-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR