Ethyl-3-fluoro-4-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-fluoro-4-nitrobenzoate

071655-1G 071655-5G 071655-10G
102520-584EA 284.62 USD
102520-584 102520-586 102520-588
Ethyl-3-fluoro-4-nitrobenzoate ≥95%
Ethyl-3-fluoro-4-nitrobenzoate

Hazard: Irritant

Formula: C₉H₈FNO₄
MW: 213.1600 g/mol
MDL Number: MFCD07782042
CAS Number: 914347-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR