2-Oxo-2-(4-phenoxyphenyl)acetaldehyde hydrate ≥95%

Supplier: Matrix Scientific

071647-1G 071647-5G 071647-10G
102520-536EA 355.38 USD
102520-536 102520-538 102520-540
2-Oxo-2-(4-phenoxyphenyl)acetaldehyde hydrate ≥95%
2-Oxo-2-(4-phenoxyphenyl)acetaldehyde hydrate

Hazard: Irritant

Formula: C₁₄H₁₂O₄
CAS Number: 92254-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR