2,2-Dihydroxy-1-(4-(piperidin-1-yl)phenyl)ethanone ≥95%

Supplier: Matrix Scientific

071646-1G 071646-5G 071646-10G
102520-530EA 355.38 USD
102520-530 102520-532 102520-534
2,2-Dihydroxy-1-(4-(piperidin-1-yl)phenyl)ethanone ≥95%

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MDL Number: MFCD05864639
CAS Number: 93290-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR