2,4,6-Trimethylphenylglyoxal hydrate ≥95%

Supplier: Matrix Scientific

071644-1G 071644-5G 071644-10G
102520-518EA 355.38 USD
102520-518 102520-520 102520-522
2,4,6-Trimethylphenylglyoxal hydrate ≥95%
2,4,6-Trimethylphenylglyoxal hydrate

Hazard: Irritant

Formula: C₁₁H₁₄O₃
CAS Number: 142751-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR