4-Acetamidophenylglyoxal hydrate ≥95%

Supplier: Matrix Scientific

Synonyms: N-(4-(2,2-Dihydroxyacetyl)phenyl)acetamide

071642-1G 071642-5G 071642-10G
102520-506EA 355.38 USD
102520-506 102520-508 102520-510
4-Acetamidophenylglyoxal hydrate ≥95%
4-Acetamidophenylglyoxal hydrate

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MW: 209.198 g/mol
MDL Number: MFCD12965019
CAS Number: 16267-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR