4-Chlorophenylglyoxal hydrate ≥95%
Supplier: Matrix Scientific
|
Danger
|
071635-1G
071635-5G
071635-10G
102520-464EA
213.85
USD
102520-464
102520-466
102520-468
4-Chlorophenylglyoxal hydrate ≥95%
4-Chlorophenylglyoxal hydrate
Purity: 95+%
CAS: 4996-21-8
MDL Number: MFCD08272327
Molecular Formula: C8H7ClO3
Molecular Weight: 186,6
Hazard: Irritant
Formula:
C₈H₅ClO₂·H₂O |
MDL Number:
MFCD08272327 CAS Number: 859932-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...