5-Fluoronicotinonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Cyano-5-fluoropyridine

071630-250MG 071630-1G
102520-442EA 936.92 USD
102520-442 102520-444
5-Fluoronicotinonitrile ≥98%
5-Fluoronicotinonitrile

Hazard: Irritant

Formula: C₆H₃FN₂
MW: 122.10 g/mol
Melting Pt: 50-54°C
MDL Number: MFCD09027295
CAS Number: 696-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR