2-Chloro-3-fluoro-4-iodopyridine ≥98%

Supplier: Matrix Scientific

071628-5G 071628-1G
102520-440EA 184.62 USD
102520-440 102833-504
2-Chloro-3-fluoro-4-iodopyridine ≥98%
2-Chloro-3-fluoro-4-iodopyridine

Hazard: Irritant

Formula: C₅H₂ClFIN
MDL Number: MFCD04972389
CAS Number: 148639-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR