4-Bromo-3-chloro-2-fluoropyridine ≥98%

Supplier: Matrix Scientific

071627-1G 071627-5G
102520-436EA 110.77 USD
102520-436 102520-438
4-Bromo-3-chloro-2-fluoropyridine ≥98%
4-Bromo-3-chloro-2-fluoropyridine

Hazard: Irritant

Formula: C₅H₂BrClFN
MW: 210.43 g/mol
MDL Number: MFCD11040607
CAS Number: 1017793-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR