α,α,α,2,5-Pentafluorotoluene ≥98%
Supplier: Matrix Scientific
Synonyms:
2,5-Difluorobenzotrifluoride
071564-1G
071564-5G
102520-342EA
112.31
USD
102520-342
102520-344
α,α,α,2,5-Pentafluorotoluene ≥98%
α,α,α,2,5-Pentafluorotoluene
Hazard: Irritant
Formula:
C₇H₃F₅ MW: 182.09 g/mol |
MDL Number:
MFCD00042494 CAS Number: 393-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...